Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I8153

Sigma-Aldrich

Anti-Interleukin-1 Receptor Accessory Protein antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IL-1RAcP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 66 kDa

species reactivity

mouse, rat, human

technique(s)

western blot: 0.5 μg/mL using human HeLa cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... IL1RAP(3556)
mouse ... Il1rap(16180)
rat ... Il1rap(25466)

General description

Interleukin-1 is a proinflammatory cytokine produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that exert their cellular effects by signaling through IL-1 receptor type 1 (IL-1R1). IL-1 recruits Interleukin-1 Receptor Accessory Protein (IL-1RAcP) that stabilizes the ligand-receptor association. IL-1RAcP is expressed by astrocytes and neurons in the brain. IL-1RAcP does not regulate the responses of IL-1 directly but modulates the gene expression and inflammatory responses induced by IL-1. IL-1RAcP is critical in mediating the activation of IRAK and SAPK pathways by IL-1. Anti-Interleukin-1 Receptor Accessory Protein antibody recognizes human, mouse and rat IL-1RAcP (~66 kDa). There is no cross-reactivity to other members in the IL-1 and IL-18 receptor families.

Immunogen

synthetic peptide corresponding to amino acids 525-540 of human Interleukin-1 Receptor Accessory Protein (Il-1RAcP). This sequence is identical in mouse and rat.

Application

Anti-Interleukin-1 Receptor Accessory Protein antibody is suitable for immunoblotting at a working concentration of 0.5 μg/ml using HeLa cell extract.

Physical form

Solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Denes et al.
Cerebrovascular diseases (Basel, Switzerland), 32(6), 517-527 (2011-11-23)
Inflammation is established as a contributor to cerebrovascular disease. Risk factors for stroke include many conditions associated with chronic or acute inflammation, and inflammatory changes in the brain after cerebrovascular events contribute to outcome in experimental studies, with growing evidence
Interleukin-1, inflammasomes, autoinflammation and the skin
Contassot E et al
Swiss Medical Weekly, 142, w13590-w13590 (2010)
H Wesche et al.
The Journal of biological chemistry, 272(12), 7727-7731 (1997-03-21)
Interleukin-1 (IL-1) is a central mediator of the immune system involved in acute and chronic inflammatory responses. Although the sequences of two types of IL-1 receptors are known, the exact molecular events resulting in signal transduction and coupling to downstream
Axel Weber et al.
Science signaling, 3(105), cm1-cm1 (2010-01-21)
The interleukin-1 (IL-1) family of cytokines comprises 11 proteins (IL-1F1 to IL-1F11) encoded by 11 distinct genes in humans and mice. IL-1-type cytokines are major mediators of innate immune reactions, and blockade of the founding members IL-1alpha or IL-1beta by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service