Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I7654

Sigma-Aldrich

Anti-Inositol 1,4,5-Triphosphate Receptor (Type II) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IP3R-II

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 240 kDa

species reactivity

rat

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The gene ITPR2 (Inositol 1,4,5-Triphosphate Receptor type 2) encodes type 2 intracellular receptor that binds to Inositol 1,4,5 trisphosphate. The gene encodes a 2701 amino acid protein that contains six membrane-spanning regions near the C-termini. The cytoplasmic N-terminal domain is involved in ligand-binding. The gene is mapped to human chromosome 12p11.

Immunogen

synthetic petide with a sequence derived from the cytoplasmic, amino-terminus of rat IP3R-II.

Biochem/physiol Actions

The receptor encoded by the gene ITPR2 (Inositol 1,4,5-Triphosphate Receptor type 2) is a Ca2+ release channel in the endoplasmic reticulum that is stimulated upon binding to its ligand inositol 1,4,5-triphosphate. It mediates intracellular calcium signaling. It facilitates the release of Ca2+ from endoplasmic reticulum and modulates intracellular Ca2+ concentration. Loss of ITPR2 gene function is found to be associated with anhidrosis and can inhibit sweat production in humans and mice.

Physical form

Solution in PBS with 1.0 mg/ml bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Futatsugi et al.
The Biochemical journal, 334 ( Pt 3), 559-563 (1998-09-08)
We have found a novel isoform of the mouse type 2 Ins(1,4,5)P3 receptor [Ins(1,4,5)P3R] mRNA by reverse transcriptase-mediated PCR analysis. The novel isoform, which was expressed specifically in skeletal muscle and heart, was generated by the inclusion of a novel
M Yamamoto-Hino et al.
Receptors & channels, 2(1), 9-22 (1994-01-01)
We have cloned cDNAs coding for human type 2 and type 3 and part of type 1 inositol 1,4,5-trisphosphate receptors (IP3Rs). The complete nucleotide sequences for type 2 and type 3 receptors were determined and the pharmacological properties of the
Joakim Klar et al.
The Journal of clinical investigation, 124(11), 4773-4780 (2014-10-21)
There are 3 major sweat-producing glands present in skin; eccrine, apocrine, and apoeccrine glands. Due to the high rate of secretion, eccrine sweating is a vital regulator of body temperature in response to thermal stress in humans; therefore, an inability

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service