Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I7388

Sigma-Aldrich

Isoxanthopterin

≥97.5%, powder

Synonym(s):

2-Amino-4,7-dihydroxy-1,3,5,8-tetraazanaphthalene, 2-Amino-4,7-dihydroxypteridine, 2-Amino-4,7-pteridinediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N5O2
CAS Number:
Molecular Weight:
179.14
Beilstein/REAXYS Number:
181594
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

assay

≥97.5%

form

powder

color

off-white

solubility

1 M NaOH: 50 mg/mL, clear, very dark yellow

storage temp.

room temp

SMILES string

NC1=NC2=C(N=CC(=O)N2)C(=O)N1

InChI

1S/C6H5N5O2/c7-6-10-4-3(5(13)11-6)8-1-2(12)9-4/h1H,(H4,7,9,10,11,12,13)

InChI key

GLKCOBIIZKYKFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isoxanthopterin is a substrate for the enzyme isoxanthopterin deaminase.

Other Notes

Product of xanthine oxidase oxidation of pterin, an enzyme involved in reactive oxygen formation.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuichi Komaki et al.
Pulmonary pharmacology & therapeutics, 18(4), 297-302 (2005-03-22)
Reactive oxygen species have been reported to be involved in the airway inflammatory process of chronic obstructive pulmonary disease (COPD). The aim of this study was to quantify the activity of xanthine oxidase (XO), which generates a potent radical superoxide
Highly sensitive assay for xanthine oxidase activity by high-performance liquid chromatography with fluorescence detection.
T Sasaoka et al.
Journal of chromatography, 424(2), 392-397 (1988-02-26)
R Walter et al.
American journal of respiratory and critical care medicine, 156(6), 2006-2010 (1997-12-31)
Pulmonary endothelial dysfunction is the hallmark of acute lung injury. Impaired pulmonary endothelial nitric oxide (NO) production in this event has been described. Tetrahydrobiopterin (BH4) is an essential cofactor for NO synthase and modulator of its activity. At high local
S Abadeh et al.
Biochimica et biophysica acta, 1117(1), 25-32 (1992-07-21)
Xanthine oxidase was purified from human milk in yields comparable with those obtained from bovine milk. The freshly purified enzyme appeared homogeneous in gel permeation FPLC and SDS-PAGE, consistent with its being a homodimer with total M(r) 290,000 +/- 6000.
Burki Rajendar et al.
Bioorganic & medicinal chemistry, 17(1), 351-359 (2008-11-18)
Isoxanthopterin (IX) has two edges with hydrogen bond-forming sites suitable for binding to thymine (T) and cytosine (C). The binding affinity of IX for T or C is stronger than for adenine (A) and guanine (G), whereas the base selectivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service