Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I6409

Sigma-Aldrich

IDR-1

≥95% (HPLC)

Synonym(s):

H-Lys-Ser-Arg-Ile-Val-Pro-Ala-Ile-Pro-Val-Ser-Leu-Leu-NH2, Innate defense regulator peptide-1, KSRIVPAIPVSLL-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H118N18O15
CAS Number:
Molecular Weight:
1391.74
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥95% (HPLC)

form

lyophilized powder (provided as TFA salt)

composition

Protein content, ~91%

solubility

H2O: soluble >1 mg/mL

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CO)NC(=O)[C@@H](N)CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N2CCC[C@H]2C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(N)=O

InChI

1S/C65H118N18O15/c1-14-37(11)50(80-55(89)41(22-18-26-71-65(69)70)73-57(91)44(31-84)76-54(88)40(67)21-16-17-25-66)62(96)79-49(36(9)10)63(97)82-27-19-23-46(82)59(93)72-39(13)53(87)81-51(38(12)15-2)64(98)83-28-20-24-47(83)60(94)78-48(35(7)8)61(95)77-45(32-85)58(92)75-43(30-34(5)6)56(90)74-42(52(68)86)29-33(3)4/h33-51,84-85H,14-32,66-67H2,1-13H3,(H2,68,86)(H,72,93)(H,73,91)(H,74,90)(H,75,92)(H,76,88)(H,77,95)(H,78,94)(H,79,96)(H,80,89)(H,81,87)(H4,69,70,71)/t37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-/m0/s1

InChI key

WXRGGPLCKMBEAF-OLAWGTLASA-N

Amino Acid Sequence

Lys-Ser-Arg-Ile-Val-Pro-Ala-Ile-Pro-Val-Ser-Leu-Leu-NH2

Biochem/physiol Actions

IDR-1 is an anti-infective peptide that acts on the host to stimulate the innate immune response.
IDR-1 is an anti-infective peptide that acts on the host to stimulate the innate immune response. IDR-1 is believed to act by enhancing monocyte chemokines, while reducing pro-inflammatory cytokines (decreased TNF and IL-6 and increased IL-10).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily Turner-Brannen et al.
Arthritis research & therapy, 13(4), R129-R129 (2011-08-13)
Innate defence regulator (IDR) peptides are synthetic cationic peptides, variants of naturally occurring innate immune effector molecules known as host defence peptides. IDR peptides were recently demonstrated to limit infection-associated inflammation selectively without compromising host innate immune functions. This study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service