Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I6402

Sigma-Aldrich

Anti-Interleukin-2 Soluble Receptor γ antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IL-2 sRγ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL2RG(3561)

General description

Interleukin-2 (IL-2) is a cytokine produced by activated T cells in response to antigen stimulation. IL-2 is important for proliferation of lymphocytes, natural killer cells and macrophages and in the differentiation of CD4+ cells into Th1 and Th2 effector cells. The effects of IL-2 are mediated by the signalling activated by its binding to IL-2 receptor (IL-2 R). IL-2 R which contains three membrane subunits: α, β and γ. The IL-2 R γ is also shared by IL-2, IL-4 and IL-7. The β and the γ subunits complex results in the binding of IL-2 with higher affinity. IL-2 R γ is expressed on monocytes, B cells, T cells, NK cells and neutrophils. IL-2 R activates key signalling pathways such as MAPK, JAK/STAT as well as PI3K/Akt. Activation of these pathways modulates production of inflammatory cytokines, survival and progression through cell cycle
Anti-Human IL-2 soluble receptor γ specifically reacts with IL-2 R γ. It does not show cross-reactivity with other cytokines.

Specificity

The antibody shows ≤2% cross-reactivity with recombinant human IL-3 sRα and IL-5 sRβ.

Immunogen

recombinant human IL-2 soluble receptor β, expressed in Sf 21 insect cells.

Application

Anti-Human IL-2 soluble receptor (IL-2 sRγ) antibody may be used in immunoblotting at a working concentration of 1-2 μg/ml. For indirect ELISA, the recommended concentration is 0.5-1.0 μg/ml. By ELISA and immunoblotting the antibody shows < 2% cross-reactivity with recombinant human IL-3 sR α and IL-5 sR. The antibody is suitable for IL-2 sRγ neutralization assays.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew A Williams et al.
Nature, 441(7095), 890-893 (2006-06-17)
Although interleukin-2 (IL-2) was initially characterized as the primary T-cell growth factor following in vitro activation, less is known about its role in shaping T-cell responses to acute infections in vivo. The use of IL-2- or IL-2-receptor-deficient mice is problematic
B H Nelson et al.
Advances in immunology, 70, 1-81 (1998-10-02)
Studies of the biology of the IL-2 receptor have played a major part in establishing several of the fundamental principles that govern our current understanding of immunology. Chief among these is the contribution made by lymphokines to regulation of the
Katrina K Hoyer et al.
Immunological reviews, 226, 19-28 (2009-01-24)
Interleukin-2 (IL-2) has multiple, sometimes opposing, functions during an inflammatory response. It is a potent inducer of T-cell proliferation and T-helper 1 (Th1) and Th2 effector T-cell differentiation and provides T cells with a long-lasting competitive advantage resulting in the
Hans Dooms et al.
Journal of immunology (Baltimore, Md. : 1950), 172(10), 5973-5979 (2004-05-07)
The acquisition of long-term survival potential by activated T lymphocytes is essential to ensure the successful development of a memory population in the competitive environment of the lymphoid system. The factors that grant competitiveness for survival to primed T cells
Arian Laurence et al.
Immunity, 26(3), 371-381 (2007-03-17)
Recent work has identified a new subset of effector T cells that produces interleukin (IL)-17 known as T helper 17 (Th17) cells, which is involved in the pathophysiology of inflammatory diseases and is thought to be developmentally related to regulatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service