The antibody will neutralize human cell surface IL-2 receptor α mediated-bioactivity. The antibody shows ≤15% cross-reactivity with recombinant human IL-2 Rγ, ≤ 5% cross-reactivity with recombinant human IL-6 sR, and ≤1% cross-reactivity with recombinant human IL-1 sRII and IL-2 sRβ.
Immunogen
recombinant human IL-2 soluble receptor α, expressed in Sf 21 insect cells.
Application
Anti-Interleukin-2 Soluble Receptor α antibody may be used in immunoblotting at a working concentration of 0.1-0.2 μg/ml. For flow cytometry, the recommended concentration is 2-3 μg/106 cells. The antibody is suitable for IL-2 sR α neutralization assays.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
The Journal of biological chemistry, 287(52), 43798-43809 (2012-10-30)
Neurotrophins comprise a group of neuronal growth factors that are essential for the development and maintenance of the nervous system. However, the immature pro-neurotrophins promote apoptosis by engaging in a complex with sortilin and the p75 neurotrophin receptor (p75(NTR)). To
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.