I6018
Sigma Lite Plus Intensifying Screens without lead blockers
For 8 × 10 in film
All Photos(1)
About This Item
UNSPSC Code:
41105341
General description
A rare earth intensifying screen for general autoradiographic use. An economical alternative to the Dupont Cronex Lightning Plus.
Choose from one of the most recent versions:
Certificates of Analysis (COA)