immunocytochemistry: 8-25 μg/mL using using human PBMCs immunohistochemistry (frozen sections): 8-25 μg/mL using using human tissue neutralization: suitable western blot: 1 μg/mL (under non-reducing conditions)
recombinant human IL-2 soluble receptor alpha (IL-2 Rα), expressed in Sf 21 insect cells.
Biochem/physiol Actions
Interleukin-2 (IL-2) is a cytokine produced by activated T cells in response to antigen stimulation. The effects of IL-2 are mediated by the signaling activated by its binding to IL-2 receptor (IL-2 R). IL-2 activates key signaling pathways such as MAPK, JAK/STAT as well as PI3K/Akt. Activation of these pathways modulates production of inflammatory cytokines, survival and progression through cell cycle.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline wth 5% trehalose.
Preparation Note
Purified using protein A.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Studies of the biology of the IL-2 receptor have played a major part in establishing several of the fundamental principles that govern our current understanding of immunology. Chief among these is the contribution made by lymphokines to regulation of the
Interleukin-2 (IL-2) has multiple, sometimes opposing, functions during an inflammatory response. It is a potent inducer of T-cell proliferation and T-helper 1 (Th1) and Th2 effector T-cell differentiation and provides T cells with a long-lasting competitive advantage resulting in the
Recent work has identified a new subset of effector T cells that produces interleukin (IL)-17 known as T helper 17 (Th17) cells, which is involved in the pathophysiology of inflammatory diseases and is thought to be developmentally related to regulatory
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.