I4895
Anti-Interleukin-9 antibody produced in goat
IgG fraction of antiserum, lyophilized powder
Synonym(s):
Anti-IL-9
About This Item
biological source
goat
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
mouse
technique(s)
neutralization: suitable
UniProt accession no.
storage temp.
−20°C
Gene Information
mouse ... Il9(16198)
General description
Anti-Interleukin-9 recognizes mouse IL-9. It does not neutralize the activity of recombinant human IL-9.
Specificity
Immunogen
Application
Physical form
Preparation Note
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service