Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I4627

Sigma-Aldrich

2′,3′-O-Isopropylidene-6-mercaptopurine riboside

Synonym(s):

2′,3′-O-Isopropylidene-6-thioinosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N4O4S
CAS Number:
Molecular Weight:
324.36
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CC1(C)O[C@H]2[C@H](CO)O[C@H]([C@H]2O1)n3cnc4c(S)ncnc34

InChI

1S/C13H16N4O4S/c1-13(2)20-8-6(3-18)19-12(9(8)21-13)17-5-16-7-10(17)14-4-15-11(7)22/h4-6,8-9,12,18H,3H2,1-2H3,(H,14,15,22)/t6?,8?,9?,12-/m1/s1

InChI key

HXZCOKXEDYVENB-LCIRSVMXSA-N

Looking for similar products? Visit Product Comparison Guide

application

2′,3′-O-Isopropylidene-6-thioinosine may be used to study metal complexing mechanisms.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal complexes of 2',3'-O-isopropylidene-6-thioinosine.
A Hampton et al.
Journal of medicinal chemistry, 9(6), 976-976 (1966-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service