Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I4532

Sigma-Aldrich

Monoclonal Anti-Interleukin-13 antibody produced in rat

clone 38213.11, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IL-13

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

38213.11, monoclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

capture ELISA: suitable
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Il13(16163)

General description

Interleukin-13 (IL-13) is a cytokine produced by Th2 cells and mediates the production of immunoglobulins, B cell proliferation and differentiation of cells of the monocytic lineage. The interaction of IL-13 with its receptor triggers responses via the STAT-6 pathway and results in antibody-mediated responses, eosinophil recruitment in lungs, airway remodelling, mucus overproduction, secretion of matrix metalloproteinases (MMP)-9 and MMP-12 and IgE production. The other signaling axis that is mediated by IL-13 is the EGFR-TGF-α pathway that induces proliferation of epithelial cells. Elevated IL-13 has been associated with the symptoms of asthma and is detected in elevated levels in the airways and sputum of asthma patients
Monoclonal Anti-Mouse Interleukin-13 recognizes recombinant mouse IL-13.

Specificity

The biological activity is measured by its ability to neutralize recombinant mouse IL-13.

Immunogen

purified recombinant mouse IL-13 expressed in E. coli.

Application

Monoclonal anti-mouse interleukin-13 may be used in neutralization assays at a working concentration range of 03.-0.8 μg/ml. For immunoblotting a concentration of 1-2 μg/ml may be used. For ELISA a working concentration of 2-4 μg/ml is recommended.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of IL-13 production by histamine in cloned murine T helper type 2 cells
Elliott, KA et al
Immunopharmacology, 1, 1923-1937 (2001)
Investigational therapeutics targeting the IL-4/IL-13/STAT-6 pathway for the treatment of asthma
Oh CK et al
European Respiratory Review, 19, 11546-11554 (2010)
Interleukin-13 and leukotrienes: an intersection of pathogenetic schema.
Jack A Elias et al.
American journal of respiratory cell and molecular biology, 28(4), 401-404 (2003-03-26)
C G Lee et al.
The Journal of experimental medicine, 194(6), 809-821 (2001-09-19)
Interleukin (IL)-13 is a key mediator of tissue fibrosis caused by T helper cell type 2 inflammation. We hypothesized that the fibrogenic effects of IL-13 are mediated by transforming growth factor (TGF)-beta. To test this hypothesis we compared the regulation
A N McKenzie et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(8), 3735-3739 (1993-04-15)
We have isolated the human cDNA homologue of a mouse helper T-cell-specific cDNA sequence, called P600, from an activated human T-cell cDNA library. The human cDNA encodes a secreted, mainly unglycosylated, protein with a relative molecular mass of approximately 10,000.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service