Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I4507

Sigma-Aldrich

2-Iminobiotin-Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

saline suspension

matrix

4% beaded agarose

matrix activation

epoxy

matrix attachment

carboxy

matrix spacer

16 atoms

capacity

≥4 mg/mL binding capacity (avidin)

storage temp.

2-8°C

Application

2-Iminobiotin-agarose is an agarose conjugate in saline suspension used in affinity chromatography, protein chromatography and avidin biotin matrices. 2-Iminobiotin-agarose has been used to improve the development of vaccines.
Interaction with avidin is pH-dependent: forms a stable complex above pH 9.5 and dissociates at pH 4.

Physical form

Suspension in 0.5 M NaCl, 0.01 M sodium phosphate, pH 6.8, containing 0.02% sodium azide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piia Karisola et al.
The Journal of biological chemistry, 277(25), 22656-22661 (2002-03-23)
A novel approach to localize and reconstruct conformational IgE-binding epitope regions of hevein (Hev b6.02), a major natural rubber latex allergen, is described. An antimicrobial protein (AMP) from the amaranth Amaranthus caudatus was used as an immunologically non-IgE-binding adaptor molecule
Complexes of Streptavidin-Fused Antigens with Biotinylated Antibodies Targeting Receptors on Dendritic Cell Surface: A Novel Tool for Induction of Specific T-Cell Immune Responses.
Stanek, O., et al.
Molecular Biotechnology (2011)
O H Laitinen et al.
FEBS letters, 461(1-2), 52-58 (1999-11-24)
Sea urchin fibropellins are epidermal growth factor homologues that harbor a C-terminal domain, similar in sequence to hen egg-white avidin and bacterial streptavidin. The fibropellin sequence was used as a conceptual template for mutation of designated conserved tryptophan residues in
Vesa P Hytönen et al.
The Biochemical journal, 384(Pt 2), 385-390 (2004-08-25)
Chicken avidin is a highly popular tool with countless applications in the life sciences. In the present study, an efficient method for producing avidin protein in the periplasmic space of Escherichia coli in the active form is described. Avidin was
M H Qureshi et al.
The Journal of biological chemistry, 276(49), 46422-46428 (2001-10-05)
The strong biotin-streptavidin interaction limits the application of streptavidin as a reversible affinity matrix for purification of biotinylated biomolecules. To address this concern, a series of single, double, and triple streptavidin muteins with different affinities to biotin were designed. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service