Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I4153

Sigma-Aldrich

Anti-Interleukin-12 Receptor β1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IL-12 Receptor β1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: 0.5-1.0 μg/mL using recombinant human IL-12 Rβ1
western blot: 0.1-0.2 μg/mL using recombinant human IL-12 Rβ1

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL12RB1(3594)

General description

IL-12 also known as Natural Killer Cell Stimulatory Factor (NKSF) is a 75kD protein that belongs to type I cytokine receptor family and is produced mainly by monocytes and NK cells. IL-12 signals with the help of IL-12 receptor made up of two subunits designated β1 and β2. Interleukin-12 receptor subunit β-1 binds interleukin-12 with low affinity and facilitates in IL12 transduction. Anti-interleukin-12 receptor β1 antibody can be used in ELISA (0.5-1.0 μg/mL) using recombinant human IL-12 Rβ1. Goat anti-interleukin-12 receptor β1 antibody has shown 5% (approx.) cross-reactivity with recombinant mouse IL-12 Rβ1.

Specificity

The antibody shows 5% cross-reactivity with recombinant mouse IL-12 Rβ1.

Immunogen

recombinant human IL-12 Rβ1 extracellular domain, expressed in NSO cells

Application

Anti-interleukin-12 receptor β1 antibody can be used in immunoblotting for localization and detection of recombinant human IL-12 Rβ1.

Physical form

Lyophilized from 0.2 μm filtered solution of phosphate buffered saline

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-12
Storkus, W.J, et al
The Cytokine Handbook, 391-425 (1998)
D H Presky et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(24), 14002-14007 (1996-11-26)
We have identified a cDNA from a human phytohemagglutinin-activated lymphoblast library encoding a protein that binds 125I-labeled human interleukin 12 (125I-huIL-12) with a Kd of about 5 nM when expressed in COS-7 cells. When coexpressed in COS-7 cells with the
A O Chua et al.
Journal of immunology (Baltimore, Md. : 1950), 153(1), 128-136 (1994-07-01)
A cDNA encoding a human IL-12R subunit was isolated by expression cloning. This subunit is a 662 amino acid type I transmembrane protein with an extracellular domain of 516 amino acids and a cytoplasmic domain of 91 amino acids. It
Lu Chen et al.
International journal of oncology, 47(4), 1361-1370 (2015-09-01)
Vasoactive intestinal peptide (VIP) has been regarded as deactivator for macrophages. However, the depressive effect of VIP on tumor-associated macrophages (TAM) has not been recognized. In the present study, we investigated the effect of VIP on gastric cancer via TAM
Christi Parham et al.
Journal of immunology (Baltimore, Md. : 1950), 168(11), 5699-5708 (2002-05-23)
IL-23 is a heterodimeric cytokine composed of the IL-12p40 "soluble receptor" subunit and a novel cytokine-like subunit related to IL-12p35, termed p19. Human and mouse IL-23 exhibit some activities similar to IL-12, but differ in their capacities to stimulate particular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service