Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I3782

Sigma-Aldrich

Insulin-like Growth Factor Binding Protein-5 from mouse

≥90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

IGFBP-5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

recombinant

expressed in NSO cells

assay

≥90% (SDS-PAGE)

form

lyophilized powder

potency

0.5-2.5 μg/mL ED50

mol wt

calculated mol wt 28.9 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Igfbp5(16011)

General description

IGFBP5 (insulin-like growth factor binding protein-5) is a secreted protein and can also move to the nucleus. IGFBPs have cysteine-rich amino- and carboxyl-terminal domains.[1]

Application

As a result of glycosylation, the recombinant mouse protein migrates to at least three bands with molecular weights ranging from 28-38 kDa in SDS-PAGE under reducing conditions.

Biochem/physiol Actions

Binds to bone cells and is the predominant insulin-like growth factor binding protein in bone; when binding the extracellular matrix, it modulates IGF activity.
IGFBP-5 binds to bone cells and is the predominant insulin-like growth factor binding protein in bone. When IGFBP-5 binds the extracellular matrix, it modulates IGF activity. Mouse, human, and rat IGFBP-5 share 97% homology.
IGFBPs (insulin-like growth factor binding proteins) control IGF functions by regulating their access to other proteins, including receptors. IGFBP5 is involved in osteogenesis, muscle differentiation and kidney development.[1] It is a crucial protein for the development of fibrosis.[2]

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% TFA containing 1.25 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to inhibit recombinant mouse IGF-II on MCF-7 cells.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidekata Yasuoka et al.
PloS one, 9(2), e87754-e87754 (2014-02-20)
Systemic sclerosis (SSc) is characterized by excessive fibrosis of the skin and internal organs due to fibroblast proliferation and excessive production of extracellular matrix (ECM). We have shown that insulin-like growth factor binding protein (IGFBP)-5 plays an important role in
Mahta Nili et al.
The Journal of biological chemistry, 287(2), 1510-1519 (2011-11-26)
The six high-affinity insulin-like growth factor-binding proteins (IGFBPs) comprise a conserved family of secreted molecules that modulate IGF actions by regulating their half-life and access to signaling receptors, and also exert biological effects that are independent of IGF binding. IGFBPs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service