Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I3157

Sigma-Aldrich

Monoclonal Anti-Interleukin-9 Receptor antibody produced in mouse

clone 33423.111, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-CD129, Anti-IL-9 Receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

33423.111, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: 0.5-1.0 μg/mL
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL9R(3581)

General description

Interleukin-9 (IL-9) is a pleotropic cytokine produced by activated T cells and also by mast cells in response to LPS and IL-1. The biological effects of IL-9 are mediated by IL-9 receptor (IL-9R) that is made up of an α and a γ chain. The expression of IL-9R is restricted to effector T cells, mast cells and few non-hematopoietic cells. IL-9R signalling results in the activation of major signalling pathways involving STAT1, -3 and -5, MAPK, PI3K and IRS. IL-9R signalling is pro-inflammatory and promotes allergic inflammation and airway remodelling in lungs. It also is important in cell growth and differentiation of T cells, B cells, mast cells, hematopoietic progenitors, epithelial cells and airway smooth muscle cells
Monoclonal Anti-Human Interleukin-9 Receptor recognizes human IL-9 R (26 kDA, observed molecular mass approximately 29 kDa as a result of glycosylation).

Specificity

The antibody blocks the cell surface human IL-9 receptor mediated biological activities induced by recombinant human IL-9.

Immunogen

purified recombinant human soluble IL-9 receptor, expressed in Sf 21 insect cells.

Application

Anti-Interleukin-9 receptor antibody may be used for immunoblotting at a working concentration of 1.0-2.0 μg/ml. For ELISA a concentration of 0.5-1.0 μg/ml is recommended. The antibody is also suitable for neutralization reactions (ND50 is 2-4 μg/ml).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ritobrata Goswami et al.
Journal of immunology (Baltimore, Md. : 1950), 186(6), 3283-3288 (2011-03-04)
IL-9 was first described in the late 1980s as a member of a growing number of cytokines that had pleiotropic functions in the immune system. Although many biological functions have been attributed to IL-9, it remains an understudied cytokine. A
Jennifer Kearley et al.
American journal of respiratory and critical care medicine, 183(7), 865-875 (2010-10-26)
IL-9 is a pleiotropic cytokine that has multiple effects on structural as well as numerous hematopoietic cells, which are central to the pathogenesis of asthma. The contribution of IL-9 to asthma pathogenesis has thus far been unclear, due to conflicting
Elizabeth C Nowak et al.
Immunology, 131(2), 169-173 (2010-08-03)
The production of interleukin-9 (IL-9) by CD4 T cells has gathered renewed interest as the result of the observation that its expression is broader than originally thought. This includes the production of IL-9 by a recently characterized subset of CD4
J C Renauld et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(12), 5690-5694 (1992-06-15)
Interleukin 9 (IL-9) is a T-cell-derived lymphokine that induces the proliferation of various lymphoid and hemopoietic cells. A cDNA clone encoding the murine IL-9 receptor was isolated by expression cloning in COS cells and screening with 125I-labeled IL-9. Transient expression
Ulla-Angela Temann et al.
The Journal of clinical investigation, 109(1), 29-39 (2002-01-10)
IL-9 is a pleiotropic cytokine with multiple functions on many cell types involved in the pathology of human asthma. The constitutive overexpression of IL-9 in the lungs of transgenic mice resulted in an asthma-like phenotype. To define the contribution of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service