Useful as a qualitative standard in protein sequencing to assure sequencer performance. Internal standard can be analyzed concurrently with sample as an internal control.
Reconstitution
Reconstitute sample in 20% acetonitrile and aliquot desired amount onto sample support.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
A sequenceable internal standard has been made that can be added to samples destined for automated amino acid sequencing and that provides a means for continuously monitoring instrument performance. The 41-residue standard peptide is composed of two unnatural amino acids
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.