I1654
Anti-IRAK2 (546-564) antibody produced in rabbit
~0.5 mg/mL, affinity isolated antibody
Synonym(s):
Anti-Interleukin-1 Receptor-Associated Kinase 2
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
mol wt
antigen 65 kDa
species reactivity
human
concentration
~0.5 mg/mL
technique(s)
western blot: 0.5-1 μg/mL using HeLa or K562 cell lysates
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... IRAK2(3656)
General description
Immunogen
Biochem/physiol Actions
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service