Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I1533

Sigma-Aldrich

Anti-phospho-Insulin Receptor Substrate-1 (pTyr941) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-phospho-IRS-1 (pTyr941)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 0.1-1.0 μg/mL using stimulated CHO cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

phosphorylation (pTyr941)

Gene Information

human ... IRS1(3667)

Immunogen

synthetic phosphopeptide derived from the region of IRS-1 phosphorylated on tyrosine 941.

Biochem/physiol Actions

Insulin receptor substrate-1 (IRS-1) is among the four IRS proteins (IRS-1-IRS-4) that mediate signalling by insulin receptor (IR) and insulin-like growth factor (IGF) receptor. IRS-1 has a pleckstrin homology domain and a phosphotyrosine-binding domain. Phosphorylation of the Tyr1179 residue in the SH-1 negatively regulates insulin. IRS-1 has the abililty to translocate to the nucleus, the mechanism of which is not very clear. Estrogen receptors (ER) α and β have a strong affinity to IRS-1, an important interaction in breast cancers and medulloblastomas. The expression of IRS-1 in prostate cancer is associated with lower tumor cell invasiveness.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 1 mg/mL BSA (IgG and protease free) and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Yang et al.
Frontiers in plant science, 7, 940-940 (2016-07-23)
Cryptochromes are photolyase-like blue light receptors that are conserved in plants and animals. Although the light-dependent catalytic mechanism of photolyase is well studied, the photochemical mechanism of cryptochromes remains largely unknown. Lack of an appropriate protein expression system to obtain
Krzysztof Reiss et al.
Journal of cellular physiology, 227(8), 2992-3000 (2012-03-29)
The family of insulin receptor substrates (IRS) consists of four proteins (IRS-1-IRS-4), which were initially characterized as typical cytosolic adaptor proteins involved in insulin receptor (IR) and insulin-like growth factor I receptor (IGF-IR) signaling. The first cloned and characterized member
X J Sun et al.
The Journal of biological chemistry, 267(31), 22662-22672 (1992-11-15)
IRS-1 is a major insulin receptor substrate which may play an important role in insulin signal transmission. The mRNA for IRS-1 in rat cells and tissues is about 9.5 kilobases (kb). Rat liver IRS-1 was stably expressed in Chinese hamster
M G Myers et al.
Endocrinology, 132(4), 1421-1430 (1993-04-01)
IRS-1 is a unique cytosolic protein that becomes tyrosine phosphorylated during insulin stimulation of intact cells and immediately associates with the phosphatidylinositol 3'-kinase (PtdIns 3'-kinase). The insulin-like growth factor-I (IGF-I) receptor also mediated the tyrosine phosphorylation of IRS-1 and increased
K Reiss et al.
Oncogene, 20(4), 490-500 (2001-04-21)
LNCaP cells are human prostatic cancer cells that have a frame-shift mutation of the tumor suppressor gene PTEN and do not express the insulin receptor substrate-1 (IRS-1), a major substrate of the type 1 insulin-like growth factor receptor (IGF-IR). Ectopic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service