Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I1282

Sigma-Aldrich

Isopentenyl triphosphate ammonium salt solution

≥95% (TLC)

Synonym(s):

3-Methylbut-2-enyl triphosphate ammonium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H13O10P3 · 4NH3
Molecular Weight:
394.19
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥95% (TLC)

packaging

vial of 200 μg

concentration

1 mg/mL in methanol: NH4OH (7:3)

shipped in

wet ice

storage temp.

−20°C

SMILES string

[NH4+].[NH4+].[NH4+].[NH4+].C\C(C)=C\COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

InChI

1S/C5H13O10P3.4H3N/c1-5(2)3-4-13-17(9,10)15-18(11,12)14-16(6,7)8;;;;/h3H,4H2,1-2H3,(H,9,10)(H,11,12)(H2,6,7,8);4*1H3

InChI key

SZFMWLMKMIAZEW-UHFFFAOYSA-N

Biochem/physiol Actions

Isoprenoid building block.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

69.8 °F

flash_point_c

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P D Wagner et al.
The Journal of biological chemistry, 275(45), 35570-35576 (2000-08-23)
The biochemical mechanism(s) by which Nm23 proteins/nucleoside diphosphate kinases suppress tumor metastasis, inhibit cell motility, and affect cellular differentiation are not known. Here we report that Nm23 proteins can phosphorylate geranyl and farnesyl pyrophosphates to give triphosphates. Wild type Nm23-H1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service