Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I1036

Sigma-Aldrich

PI3K (p110β/p85α), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp779K1237, GRB1, MGC133043, PIK3C1, PIK3CB, PIK3R1, p110β, p85α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

13-19 nmol/min·mg

mol wt

p110β ~111 kDa
p110β/p85α ~85 kDa

UniProt accession no.

relevant disease(s)

cancer (prostate tumor)

shipped in

dry ice

Storage temp.

−70°C

Gene Information

Biochem/physiol Actions

The PI3K (phosphatidylinositol-4,5-bisphosphate 3 kinase) comprises of a 110kDa catalytic subunit and a 85kDa regulatory subunit.[1] The p110β catalytic subunit (PIK3Cβ, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit β) controls the formation and stability of αIIbβ3 integrin adhesion bonds. This event is needed for shear force-mediated platelet activation.[2] Absence of p110β impedes tumorigenesis in animals having prostate tumor, due to loss of PTEN (phosphatase and tensin homolog).[3]

Physical form

Supplied in 50 mM Sodium Phosphate, pH 7.0, with 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaun P Jackson et al.
Nature medicine, 11(5), 507-514 (2005-04-19)
Platelet activation at sites of vascular injury is essential for the arrest of bleeding; however, excessive platelet accumulation at regions of atherosclerotic plaque rupture can result in the development of arterial thrombi, precipitating diseases such as acute myocardial infarction and
Mattia Barbareschi et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 13(20), 6064-6069 (2007-10-20)
In breast cancer, the PIK3CA gene is frequently mutated at "hotspots" in exons 9 and 20, corresponding to the helical and kinase domains, respectively. We decided to investigate the association of PIK3CA mutations with pathologic features and clinical outcome in
Shidong Jia et al.
Nature, 454(7205), 776-779 (2008-07-03)
On activation by receptors, the ubiquitously expressed class IA isoforms (p110alpha and p110beta) of phosphatidylinositol-3-OH kinase (PI(3)K) generate lipid second messengers, which initiate multiple signal transduction cascades. Recent studies have demonstrated specific functions for p110alpha in growth factor and insulin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service