I1029
Interleukin-2 Soluble Receptor γ human
>97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder
Synonym(s):
IL-2 sRγ
About This Item
biological source
human
recombinant
expressed in baculovirus infected Sf21 cells
assay
>97% (SDS-PAGE)
form
lyophilized powder
potency
1.979 μg/mL ED50
mol wt
40 kDa
packaging
pkg of 50 μg
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... IL2RG(3561)
General description
Biochem/physiol Actions
Physical form
Analysis Note
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service