Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I0529

Sigma-Aldrich

Anti-Interleukin-16 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IL-16

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: 0.5-1.0 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL16(3603)

General description

Interleukin-16 is also known as lymphocyte chemoattractant factor (LCF) and acts as chemoattractant for human T cells. IL-16 is secreted by cells of immune system such as T cells, dendritic cells and eosinophils. It is also secreted by the non-immune cells such as endothelial cells, fibroblasts and neurons. IL-16 is biologically active in the multimeric form and binds to CD4 and CCR5 receptors. The signaling induced by IL-16 activates pathways that involve MAPK, PI3K and SAPK. The biological activities of IL-16 are important for inflammation, migration, cell cycle progression, cell proliferation, development of Th1 cells and regulation of T cell responses. IL-16 signaling is important in the pathophysiology of Crohn′s disease, HIV infection, airway inflammation in asthma and rheumatoid arthritis. Anti-Interleukin-16 antibody recognizes human IL-16.

Immunogen

recombinant human IL-16 expressed in E. coli.

Application

Anti-Interleukin-16 antibody may be used for immunoblotting at a working antibody concentration of 0.1-0.2 μg/ml. For ELISA, a working concentration of 0.5-1.0 μg/ml is recommended.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-16 in asthma.
Jing-min Deng et al.
Chinese medical journal, 119(12), 1017-1025 (2006-06-30)
W W Cruikshank et al.
Journal of leukocyte biology, 67(6), 757-766 (2000-06-17)
Interleukin 16 (IL-16) was initially described in 1982 as the first T cell chemoattractant. Through interaction with CD4, IL-16 has now been characterized as a chemoattractant for a variety of CD4+ immune cells. Recent in vivo studies have more fully
Monocytes or T cells in Crohn's disease: does IL-16 allow both to play at that game?
S Schreiber
Gut, 49(6), 747-748 (2001-11-16)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service