Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HUTR09854

Sigma-Aldrich

MISSION® 3′UTR Lenti GoClone

Powered by SwitchGear Genomics, 3′UTR, human, SFTPB

Synonym(s):

HUTR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

description

Functional Validation of miRNA Gene Targets

product line

MISSION®

concentration

≥1x105 VP/ml (via p24 assay)

human 3′UTR sequence

TTCCAGCTTTTGATTGAAAGTCCTAGGGTGATTCTATTTCTGCTGTGATTTATCTGCTGAAAGCTCAGCTGGGGTTGTGCAAGCTAGGGACCCATTCCTGTGTAATACAATGTCTGCACCAATGCTAATAAAGTCCTATTCTCTTTTATGAGAAAGAAAAAGACACCGTCCTTTAAAGTGCTGCAGTATggccagacgtggtggctcacacctgcaatcccagcaccttaggaggccgaggcaggaggatccttg (3′UTR sequences are truncated after the first 255 bases)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SFTPB(6439)

Application

To see more application data, protocols, and vector maps visit www.sigma.com/3utr.

Physical form

200 μL of at least 105 VP/mL (via p24 titering assay) lentiviral particles are provided as frozen stock.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see www.sigmaaldrich.com/missionlicense.
GoClone is a trademark of SwitchGear Genomics
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
SwitchGear Genomics is a trademark of SwitchGear Genomics

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walaa A Abuelhamed et al.
Indian pediatrics, 52(5), 391-394 (2015-06-11)
To study the relationship between SP-A2 and SP-B gene polymorphisms and respiratory distress syndrome in preterm neonates. Cross-sectional. Neonatal intensive care unit and the Molecular Biology unit of the Chemical Pathology Department, Kasr Alainy hospital, Cairo University. Sixty-five preterm infants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service