HUTR05654
MISSION® 3′UTR Lenti GoClone™
Powered by SwitchGear Genomics™, 3′UTR, human, SUV420H2
Synonym(s):
HUTR
About This Item
description
Functional Validation of miRNA Gene Targets
product line
MISSION®
concentration
≥1x105 VP/ml (via p24 assay)
human 3′UTR sequence
GGATGTCGGCGGTGAAGAGCTGTGACAGGCCGGACGGGGAGGCCCAGCAGGGAGAGAGGGTCTCTCTCCTAGCTGCTACCCAGGACCTCCAGAAGGAGCCCTTGGACCTCTGGGAGGGAGCTGACCCTTGACTCCAGCATAGCTCTGACCCTGGAATGGGGTTGGTTTGGACACCCCCAGGGATCTGAGCCCTGACCCTTTGTGACTGCTGACCCCTGAGCCACCCCCACTCCCACAGGGAGCCCCGGCCATTTG (3′UTR sequences are truncated after the first 255 bases)
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... SUV420H2(84787)
Application
Physical form
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service