Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HSTUD0346

Sigma-Aldrich

MISSION® Synthetic microRNA Inhibitor, Human

hsa-miR-19b-3p

Synonym(s):

hsa-miR-19b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

product line

MISSION®

form

solid

mature sequence

UGUGCAAAUCCAUGCAAAACUGA

Sanger mature/minor accession no.

Sanger microRNA accession no.

storage temp.

−20°C

General description

Individual synthetic microRNA inhibitors were designed using a proprietary algorithm, which is based on the work of Haraguchi, T, et al. and in collaboration with Dr. Hideo Iba, University of Tokyo. This algorithm utilizes the tough decoy (TuD) design. miRNA are known to regulate gene expression in a variety of manners, including translational repression, mRNA cleavage and deadenylation.

The MISSION synthetic miRNA Inhibitors are small, double-stranded RNA molecules designed to inhibit a specific mature miRNA. The miRNA inhibitors were designed using the mature miRNA sequence information from miRBase and are 2′-O-methylated RNA duplexes with a miRNA binding site on each strand. Optimal miRNA inhibition is provided after transfection due to the robust secondary structure of the inhibitor.

  • Long lasting inhibition at very low dosage
  • Excellent resistance to cellular nucleases
  • Custom synthesis available for a variety of species

Two negative controls are available: Arabidopsis thaliana sequence (NCSTUD001) and Caenorhabditis elegans sequence (NCSTUD002)

Other Notes

Based on miRBase V19

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT RE 2 Inhalation

target_organs

Respiratory Tract

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service