HS28624
TrueNorth® Upright Freezer Rack
for multi-well assay plates, 4 x 4 configuration, stainless steel
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122806
NACRES:
NB.22
Recommended Products
material
stainless steel
feature
4 x 4 configuration
Holds 16 x plates
manufacturer/tradename
Heathrow Scientific HS28624
technique(s)
cryopreservation: suitable
L × W × H
21.3 × 4.4 × 9 in.
size
543x112x23 mm
Looking for similar products? Visit Product Comparison Guide
General description
- TrueNorth® Upright Freezer Racks are ideal for safely storing the samples.
- Choose from a wide selection of corrosion-resistant stainless-steel racks that are compatible with major brands of upright and chest freezers.
- Features handle at each end accomodate freezers with right or left handed doors for easy use.
- Compatible with 6 in. (15.2 cm), or smaller boxes
Choose from a wide selection of corrosion resistant stainless steel racks that are compatible with major brands of upright and chest freezers.
All have handles for easy use.
All have handles for easy use.
Other Notes
- holds 48 ea/3 per compartment of 96-well assay plates
- holds 32 ea/2 per compartment of 24-well assay plates
- holds 16 ea/1 per compartment of 96-deep well assay plates
Legal Information
TrueNorth is a registered trademark of Heathrow Scientific LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service