Key Documents
HS24780
Lab Alert® big digit timer and clock
About This Item
UNSPSC Code:
41122411
NACRES:
NB.51
manufacturer/tradename
Heathrow Scientific HS24780
General description
Big-Digit Dual Clock/Timer
  • Display clock and timer simultaneously, or two independent timers
  • Use one timer to count up, or use two timers to count down independently
  • Audible and visual alarm
  • 142 x 97 x 20 mm
Legal Information
Lab Alert is a registered trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1