HS120357
TracRack® Storage Rack
Micro, holds 64 x 0.5 mL or 96 x 0.2 mL tubes, polypropylene
About This Item
Recommended Products
material
polypropylene
feature
Holds 64 x 0.5 Ml
Holds 96 x 0.2 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120357
technique(s)
PCR: suitable
well diam. × diam.
5.1 mm × 7.8 mm
Looking for similar products? Visit Product Comparison Guide
General description
Eight individual segments are nested in a 2" polypropylene film box that allows users to easily open, identify and remove only the segments of samples needed.
Once the segments are removed, simply replace the box lid and leave the remaining samples in storage.
Each tube segment is made from durable polypropylene and allows for easy working while saving valuable counter space.
Two versions are available, tracrack that accomodates 1.5-2 mL cyro and micro centrifuge tubes, and the reversible tracrack micro which will hold 64 x 0.5 mL microtubes on one side and 96 x 0.2 mL micro tubes on the other.
Features and Benefits
- segments made from polypropylene
- eight segments per box
- outer box is made from thin film polypropylene material
- store down to temperatures of -80C
- moisture resistant, significantly reducing the change of mold
- removable segments allow outer shell to be used for storage of other small laboratory products
- light tight box for light sensitive materials
- 2" box fits in standard freezer racks
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service