Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HR-0760000

Sigma-Aldrich

Piv2TET-Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H26Cl4O10
Molecular Weight:
712.35
UNSPSC Code:
41121815
Pricing and availability is not currently available.

product line

Proligo Reagents

assay

≥97.0% (reversed phase HPLC)

form

solid

impurities

≤3.0% 5-isomere (reversed phase HPLC)

color

white to yellow

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR

storage temp.

room temp

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service