Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA047185

Sigma-Aldrich

Anti-PIEZO1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FAM38A, Anti-KIAA0233, Anti-piezo-type mechanosensitive ion channel component 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20-1:50

immunogen sequence

SVVGVVNQPIDVTVTLKLGGYEPLFTMSAQQPSIIPFTAQAYEELSRQFDPQPLAMQFISQYSPEDIVTAQIEGSSGALWRISPPSRAQMKRELYNGTADITLRFTWNFQRDLAKGGTVEYANEKH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIEZO1(9780)

General description

Piezo type mechanosensitive ion channel component 1 (PIEZO1), also known as fam38a, is encoded by the gene mapped to human chromosome 16q24.3. The protein contains 30 transmembrane domains and is expressed in variety of tissues, such as skin, kidney, lung and colon.

Immunogen

piezo-type mechanosensitive ion channel component 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Piezo type mechanosensitive ion channel component 1 (PIEZO1) facilitates fluid shear stress response of the vascular endothelium. It is also essential for maintaining vascular structure and embryonic development. Piezo1 is involved in the lineage choice of stem cell to neuron/astrocyte differentiation. In addition, it also aids in sensing nanoroughness of the environment. Piezo1 plays a vital role in maintaining erythrocyte volume homeostasis. Aberrations in the expression of the gene lead to swelling/lysis of red blood cells and consequent anemia. Mutation in the gene is associated with the development of hereditary xerocytosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71755

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human PIEZO1 ion channel functions as a split protein.
Bae C, et al.
PLoS ONE, 11(3), e0151289-e0151289 (2016)
Piezo1 plays a role in erythrocyte volume homeostasis.
Faucherre A, et al.
Haematologica (2013)
Piezo1 integration of vascular architecture with physiological force.
Li J, et al.
Nature, 515(7526), 279-279 (2014)
The biophysics of piezo1 and piezo2 mechanosensitive channels.
Soattin L, et al.
Biophysical Chemistry, 208, 26-33 (2016)
Novel mechanisms of PIEZO1 dysfunction in hereditary xerocytosis.
Glogowska E, et al.
Blood (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service