Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA044079

Sigma-Aldrich

Anti-A1CF antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ACF, Anti-ACF64, Anti-ACF65, Anti-APOBEC1CF, Anti-ASP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

REIYMNVPVGAAGVRGLGGRGYLAYTGLGRGYQVKGDKREDKLYDILPGMELTPMNPVTLKPQGIKLAPQILEEICQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... A1CF(29974)

General description

A1CF (apobec-1 complementation factor) codes for apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1 (APOBEC1). It is a member of the heterogeneous nuclear ribonucleoproteins (hnRNP) family. A1CF has three non-identical RNA recognition motifs (RRM) and a unique C-terminal auxiliary domain.

Immunogen

APOBEC1 complementation factor

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

A1CF (apobec-1 complementation factor) modulates expression of the gene by regulating the constancy of mRNA and RNA editing. It plays a vital role in the synthesis of ApoB (both isoforms ApoB-48 and ApoB-100). Overexpression of APOBEC-1 in transgenic animals results in liver dysplasia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST80274

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of the GCKR and A1CF loci with alcohol consumption to influence the risk of gout.
Rasheed H, et al.
Arthritis Research & Therapy, 19(1), 161-161 (2017)
Identification of a novel variant of the RET proto-oncogene in a novel family with Hirschsprung?s disease.
Kawano T, et al.
Pediatric Surgery International, 33(10), 1041-1046 (2017)
Apobec-1 complementation factor regulates cell migration and apoptosis through Dickkopf1 by acting on its 3? untranslated region in MCF7 cells.
Yan X, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(6), 1010428317706218-1010428317706218 (2017)
A Chester et al.
Biochimica et biophysica acta, 1494(1-2), 1-13 (2000-11-10)
RNA editing is a post-transcriptional process that changes the informational capacity within the RNA. These processes include alterations made by nucleotide deletion, insertion and base conversion. A to I and C to U conversion occurs in mammals and these editing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service