Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA042720

Sigma-Aldrich

Anti-VKORC1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-VKCFD2, Anti-Vitamin K Epoxide Reductase Complex, Subunit 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ARDRDYRALCDVGTAISCSRVFSSRW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VKORC1(79001)

General description

Vitamin K epoxide reductase complex subunit 1 (VKORC1), also known as vitamin K clotting factor deficiency type 2 (VKCFD2), is an integral, 163-amino acid long transmembrane protein. It is encoded by the gene mapped to a 20Mb region on human chromosome 16p12–q21. VKORC1 is widely expressed in various tissues and organs.

Immunogen

vitamin K epoxide reductase complex, subunit 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Vitamin K epoxide reductase complex subunit 1 (VKORC1) enzyme plays a vital role in vitamin K cycle. It catalyzes the conversion of inactive vitamin K to vitamin K hydroquinone, which is crucial for the post-translational Γ-carboxylation of vitamin K–dependent clotting factors II (prothrombin), VII, IX, and X. VKORC1 acts as a key target for oral anticoagulants and is also involved in drug metabolism. Genetic variation in the gene leads to warfarin resistance and multiple coagulation factor deficiency type 2.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83674

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in VKORC1 cause warfarin resistance and multiple coagulation factor deficiency type 2.
Rost S
Nature null
G-1639A but Not C1173T VKORC1 Gene Polymorphism Is Related to Ischemic Stroke and Its Various Risk Factors in Ukrainian Population.
Dubovyk YI
BioMed Research International null
High prevalence of VKORC1*3 (G9041A) genetic polymorphism in north Indians: A study on patients with cardiac disorders on acenocoumarol.
Sehgal T
Drug Discoveries & Therapeutics null
VKCFD2 - from clinical phenotype to molecular mechanism.
Czogalla KJ
Hamostaseologie null
Massimo Alfano et al.
Fertility and sterility, 111(4), 687-698 (2019-04-02)
To study pathogenic features of the somatic testicular microenvironment associated with idiopathic germ cell aplasia. Cross-sectional study. Tertiary referral center for reproductive medicine. Testicular specimens from men with idiopathic nonobstructive azoospermia (iNOA) prospectively submitted to microdissection testicular sperm extraction. Of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service