Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA040616

Sigma-Aldrich

Anti-PIEZO2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C18orf30, Anti-C18orf58, Anti-FAM38B, Anti-FAM38B2, Anti-FLJ23144, Anti-FLJ23403, Anti-FLJ34907, Anti-HsT748, Anti-HsT771, Anti-piezo-type mechanosensitive ion channel component 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

FEDENKAAVRIMAGDNVEICMNLDAASFSQHNP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIEZO2(63895)

General description

Piezo-type mechanosensitive ion channel component 2 (PIEZO2) is encoded by the mapped to human chromosome 18p. It is mainly expressed in the urinary bladder, lung, dorsal root ganglia and in Merkel cells in the skin.

Immunogen

piezo-type mechanosensitive ion channel component 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Piezo-type mechanosensitive ion channel component 2 (PIEZO2) functions as a non-selective cation channel. The encoded protein is a mechanosensitive ion channel and is involved in light-touch mechanosensation. PIEZO2 is also a mechanotransduction channel for proprioception. Dominant mutations causing gain of function in PIEZO2 result in Gordon syndrome/distal arthrogryposis and Marden-Walker syndrome. Recessive mutations in this gene lead to distal arthrogryposis with distal muscle weakness, scoliosis and proprioception problems.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81707.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frédéric Lanthier et al.
IBRO neuroscience reports, 15, 42-49 (2023-07-07)
Marsupials are born very immature yet must be sufficiently autonomous to crawl on the mother's belly, find a teat and attach to it to pursue their development. Sensory inputs are necessary to guide the newborn to a teat and induce
Mosaicism of tetrasomy 18p: clinical and cytogenetic findings in a female child.
Bai J L, et al.
Chinese Medical Journal (English Edition), 130(6), 744-744 (2017)
Raquel Martín-Sanz et al.
Cancers, 16(13) (2024-07-13)
In the last decade, a group of Ca2+ channels called Piezo were discovered, demonstrating a decisive role in the cellular response to mechanical stimuli and being essential in the biological behavior of cells regarding the extracellular compartment. Several investigations have
Biallelic loss of proprioception-related PIEZO2 causes muscular atrophy with perinatal respiratory distress, arthrogryposis, and scoliosis.
Delle Vedove A, et al.
American Journal of Human Genetics, 99(5), 1206-1216 (2016)
Recessive PIEZO2 stop mutation causes distal arthrogryposis with distal muscle weakness, scoliosis and proprioception defects.
Haliloglu G, et al.
Journal of Human Genetics, 62(4), 497-497 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service