Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA040355

Sigma-Aldrich

Anti-DGKH antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dgketa, Anti-Diacylglycerol kinase, eta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

NKANPRCPESLTRDTATEIAINVKALYNETESLLVGRVPLQLESPHEERVSNALHSVEVELQKLTEIPWLYYILHPNEDEEPPMDCTKRNNRS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DGKH(160851)

General description

Diacylglycerol kinase η (DGKH) is encoded by the gene mapped to human chromosome 13q14. The encoded protein belongs to the type II diacylglycerol kinase (DGK) enzyme family. DGKH has two alternatively spliced transcript variants encoding different isoforms namely, η1 and η2. Both the isoforms contain catalytic a and b subdomains and N-terminal pleckstrin homology domain (PH) in common. DGKH η2 isoform has sterile α-motif domain at its C terminal end, whereas it is absent in η1 isoform.

Immunogen

diacylglycerol kinase, eta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Diacylglycerol kinase η (DGKH) is a vital constituent of lithium-sensitive phosphatidyl inositol pathway. The encoded protein plays an essential role in the regulation of intracellular concentrations of two bioactive lipids, including diacylglycerol and phosphatidic acid. DGKH functions as a potential cellular sensor for phosphatidylinositol (PI) 4,5-bisphosphate (PI(4,5)P2). Mutations in the gene are associated with bipolar disorder (BPD), adult attention-deficit/hyperactivity disorder (aADHD) and unipolar depression (UPD). In addition, variations in the gene expression also increase the risk of susceptibility to calcium oxalate stone and hypercalciuria.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81294

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Baum et al.
Molecular psychiatry, 13(2), 197-207 (2007-05-09)
The genetic basis of bipolar disorder has long been thought to be complex, with the potential involvement of multiple genes, but methods to analyze populations with respect to this complexity have only recently become available. We have carried out a
Aiko Kume et al.
The Journal of biological chemistry, 291(15), 8150-8161 (2016-02-19)
Type II diacylglycerol kinase (DGK) isozymes (δ, η, and κ) have a pleckstrin homology domain (PH) at their N termini. Here, we investigated the lipid binding properties of the PHs of type II DGK isozymes using protein-lipid overlay and liposome
Yong Xu et al.
Urolithiasis, 42(5), 379-385 (2014-08-02)
Diacylglycerol kinase eta (DGKH) participates in regulating the intracellular concentrations of two bioactive lipids, diacylglycerol and phosphatidic acid. With the emerging evidence of a novel regulatory function for diacylglycerol kinase and diacylglycerol in transplasmalemmal calcium ion influx, the present study
Heike Weber et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 36(10), 2076-2085 (2011-06-10)
Recently, several genome-wide association studies (GWAS) on bipolar disorder (BPD) suggested novel risk genes. However, only few of them were followed up and further, the specificity of these genes is even more elusive. To address these issues, we genotyped SNPs
Victoria B Bartsch et al.
Genes, brain, and behavior, 19(3), e12570-e12570 (2019-04-16)
Genome-wide association studies linked diacylglycerol kinase eta and iota to mood disorders, including bipolar disorder and schizophrenia, and both genes are expressed throughout the brain. Here, we generated and behaviorally characterized female mice lacking Dgkh alone, Dgki alone, and double

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service