Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA039526

Sigma-Aldrich

Anti-ATP12A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ATP1AL1, Anti-ATPase, H+/K+ transporting, nongastric, α polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.4 μg/mL
immunohistochemistry: 1:200- 1:500

immunogen sequence

IYSVELSGTKDIVKTDKGDGKEKYRGLKNNCLELKKKNHKEEFQKELHLDDHKLSNRELEEKYGTDIIMGLSSTR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP12A(479)

General description

The ATPase H+/K+ transporting non-gastricα2 subunit (ATP12A) gene, with 23 exons and 22 introns spanning 32kb of genomic DNA, is mapped to human chromosome 13q12.1-q12.3. ATP12A is a member of P-type cation transport ATPases family. The gene encodes the catalytic alpha subunit of Na+/K+-ATPase, which is ubiquitously expressed.

Immunogen

ATPase, H+/K+ transporting, nongastric, alpha polypeptide recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper) and immunofluorescence.

Biochem/physiol Actions

ATPase H+/K+ transporting non-gastric α2 subunit (ATP12A) plays a vital role in the regulation of acid-base and K+ homeostasis in the kidney and colon. In addition, it also aids acidification of prostate secretions in the anterior lobe of the organ and transepithelial bicarbonate secretion in the ducts of the exocrine pancreas. ATP12A mutations might affect myocardial Na+ handling and subsequently myocardial function. Overexpression of ATP12A has been observed in human colorectal carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81283.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Left ventricular diastolic function associated with common genetic variation in ATP12A in a general population.
Knez J
BMC Medical Genetics (2014)
The putative role of the non-gastric H?/K?-ATPase ATP12A (ATP1AL1) as anti-apoptotic ion transporter: effect of the H?/K? ATPase inhibitor SCH28080 on butyrate-stimulated myelomonocytic HL-60 cells.
Jakab M
Cellular Physiology and Biochemistry, 34, 1507-1526 (2014)
Goblet Cell Hyperplasia Requires High Bicarbonate Transport To Support Mucin Release.
Gorrieri G
Scientific Reports (2016)
Expression of the non-gastric H+/K+ ATPase ATP12A in normal and pathological human prostate tissue.
Streif D
Cellular Physiology and Biochemistry, 28, 1287-1294 (2011)
Giulia Gorrieri et al.
Scientific reports, 6, 36016-36016 (2016-10-28)
Goblet cell hyperplasia, a feature of asthma and other respiratory diseases, is driven by the Th-2 cytokines IL-4 and IL-13. In human bronchial epithelial cells, we find that IL-4 induces the expression of many genes coding for ion channels and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service