Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA035831

Sigma-Aldrich

Anti-CDC7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDC7L1, Anti-HsCdc7, Anti-Hsk1, Anti-huCdc7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ATAQLQVGPEEKIALKHLIPTSHPIRIAAELQCLTVAGGQDNVMGVKYCFRKNDHVVIAMPYLEHESFLDILNSLSFQEVREY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDC7(8317)

General description

Cell division cycle 7 kinase (CDC7), a serine/threonine kinase, comprises two or three kinase insert domains and 11 kinase domains. It has a canonical bilobal kinase fold in its structure. The CDC7 gene is localized on human chromosome 1p22.2.

Immunogen

cell division cycle 7

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cell division cycle 7 kinase (CDC7) mediates the phosphorylation of the minichromosome maintenance protein 2 (MCM2). Elevated levels of Cdc7 are observed in tumors. The inhibition of Cdc7 suppresses invasiveness and tumor growth in glioblastoma. The presence of dumbbell forming factor 4 (DBF4) is crucial for Cdc7 kinase activity. Cdc7 is an S phase-promoting kinase and favors DNA replication Cdc7 is also involved in chromosome separation and transactions. It inhibits collagen uptake and its silencing results in an upsurge of collagen uptake.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79588

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryo Kitamura et al.
The Journal of biological chemistry, 286(26), 23031-23043 (2011-05-04)
Cdc7 is a serine/threonine kinase conserved from yeasts to human and is known to play a key role in the regulation of initiation at each replication origin. Its catalytic function is activated via association with the activation subunit Dbf4/activator of
Michael J Podolsky et al.
American journal of physiology. Lung cellular and molecular physiology, 315(3), L360-L370 (2018-05-25)
Although extensive work has delineated many of the mechanisms of extracellular matrix (ECM) production, far less is known about pathways that regulate ECM degradation. This is particularly true of cellular internalization and degradation of matrix, which play an underappreciated role
Samual D Dick et al.
Structure (London, England : 1993), 28(8), 954-962 (2020-06-11)
CDC7 is an essential Ser/Thr kinase that acts upon the replicative helicase throughout the S phase of the cell cycle and is activated by DBF4. Here, we present crystal structures of a highly active human CDC7-DBF4 construct. The structures reveal
Dorine Bonte et al.
Neoplasia (New York, N.Y.), 10(9), 920-931 (2008-08-21)
Cdc7 is a conserved serine/threonine kinase essential for the initiation of DNA replication, likely by activating the MCM DNA helicase at the G(1-) to S-phase transition. Cdc7 kinase activity requires association with its regulatory subunit Dbf4/activator of S-phase kinase. Cdc7-Dbf4
Zubeyde Erbayraktar et al.
Cancer cell international, 16, 88-88 (2016-11-29)
Genomic instability is a hallmark of cancer cells, and this cellular phenomenon can emerge as a result of replicative stress. It is possible to take advantage of replicative stress, and enhance it in a targeted way to fight cancer cells. One

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service