Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA031074

Sigma-Aldrich

Anti-CTGF antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1

Synonym(s):

Ctgf Antibody, Ctgf Antibody - Anti-CTGF antibody produced in rabbit, Anti-CCN2, Anti-IGFBP8, Anti-connective tissue growth factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: None
immunofluorescence: 1-4 μg/mL
immunohistochemistry: 1:20- 1:50

immunogen sequence

CTERDPCDPHKGLFCDFGSPANRKIGVCTAKDGAPCIFGGTVYRSGESFQSSCKYQCTCLDGAVGCMPLC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTGF(1490)

General description

The CTGF (connective tissue growth factor) gene is mapped to human chromosome 6q23.2. The encoded protein contains five domains namely, secretory signal peptide, insulin-like growth factor-binding protein, von Willebrand factor type C repeat, thrombospondin type-1 repeat and C-terminal cystine-knot-containing domain. Th gene is broadly expressed in many cells including fibroblasts, myofibroblasts, endothelial cells, smooth muscle cells and some epithelial cell types. It is either released to extracellular matrix or is located on the cell membrane.

Immunogen

connective tissue growth factor recombinant protein epitope signature tag (PrEST)

Application

Anti-CTGF antibody produced in rabbit has been used in immunohistochemistry. [1]
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

CTGF (connective tissue growth factor) regulates cell communication with the extracellular matrix also inducing collagen deposition, stimulation and differentiation of mesenchymal cells and tissue remodeling. Abnormal expression of CTGF is associated with pathological conditions such as arthritis, fibrosis, and cancers. CTGF might control epithelial-mesenchymal transition, a signal pathway that contains an important step associated with tumor cell metastasis, in many types of cancer. Overexpression of CTGF enhances esophageal squamous cell carcinoma by associating with TGF-β (transforming growth factor β) pathway. It is involved in the development of fibrotic tissues in different types of organs. Overexpression of CTGF leads to fibrosis in many disease condition. Overexpression of CTGF is observed in joint capsule during joint contracture. CTGF is associated with inflammation reactions and controls macrophage adhesion and migration.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77407

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased Epithelial Expression of CTGF and S100A7 with Elevated Subepithelial Expression of IL-1? in Trachomatous Trichiasis.
Derrick T
PLoS Neglected Tropical Diseases, 10(6) (2016)
The Hippo transducers TAZ/YAP and their target CTGF in male breast cancer.
Di Benedetto A
Oncotarget, 7(28), 43188-43198 (2016)
Anna Di Benedetto et al.
Oncotarget, 7(28), 43188-43198 (2016-06-02)
Male breast cancer (MBC) is a rare disease and its biology is poorly understood. Deregulated Hippo pathway promotes oncogenic functions in female breast cancer. We herein investigated the expression of the Hippo transducers TAZ/YAP and their target CTGF in MBC.
Decreased expression of connective tissue growth factor in non-small cell lung cancer is associated with clinicopathological variables and can be restored by epigenetic modifiers.
Drzewiecka H
Journal of Cancer Research and Clinical Oncology, 142(9), 1927-1946 (2016)
MicroRNA-145 Inhibits Cell Migration and Invasion and Regulates Epithelial-Mesenchymal Transition (EMT) by Targeting Connective Tissue Growth Factor (CTGF) in Esophageal Squamous Cell Carcinoma.
Han Q
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 22, 3925-3934 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service