Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA031065

Sigma-Aldrich

Anti-NPR3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-19A, Anti-ANPRC, Anti-CD319, Anti-CRACC, Anti-CS1, Anti-GUCY2B, Anti-NPRC, Anti-natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

GFQHKDSEYSHLTRVAPAYAKMGEMMLALFRHHHWSRAALVYSDDKLERNCYFTLEGVHEVFQEEGLHTSIYSFDETKDLDLEDIVRNIQASERVVIMCASSDTIRSIMLVAHRHGMTSGDYAFFNIELFNSSSYGDGSW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPR3(4883)

General description

Natriuretic peptide receptor 3 (NPR3) or natriuretic peptide receptor type C (NPR-C) protein is widely expressed in lungs, kidneys, endocrine glands, endothelial cells, and vessels. This protein comprises an extracellular domain, a transmembrane domain, and a short intracellular domain. NPR3 gene is located on the human chromosome at 5p13.3.

Immunogen

natriuretic peptide receptor 3

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-NPR3 antibody produced in rabbit has been used in immunohistochemistry(1:30)[1], (1:25).

Biochem/physiol Actions

Natriuretic peptide receptor 3 (NPR3) or natriuretic peptide receptor type C (NPR-C) is a scavenger receptor that mediates clearance of natriuretic peptides. This protein also plays a role in inhibiting the adenylate cyclase and activates phospholipase C. NPR3 protein lacks intrinsic enzymatic activity. This protein is involved in the pathogenesis of atherosclerosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85103

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Hu et al.
Oncotarget, 7(23), 33662-33674 (2016-05-19)
To find a new locus that confers significant susceptibility to CAD in Chinese Han population, a genome-wide association study in 200 "extreme individuals" from a Shandong cohort and a pathway-based candidate gene study from a Shanghai cohort (293 CAD/293 controls)
Celia L Gregson et al.
Bone, 114, 62-71 (2018-06-09)
Generalised high bone mass (HBM), associated with features of a mild skeletal dysplasia, has a prevalence of 0.18% in a UK DXA-scanned adult population. We hypothesized that the genetic component of extreme HBM includes contributions from common variants of small
D-B Kuang et al.
Journal of clinical pharmacy and therapeutics, 42(5), 554-560 (2017-05-13)
Essential hypertension (EH) is a common disease exhibiting large individual difference in occurrence, development and treatment response. Genetic factors are implicated in the development and progression of EH. This study aimed to explore the association between NPR3 single nucleotide polymorphism
Kavneet Kaur et al.
Brain pathology (Zurich, Switzerland), 26(3), 334-343 (2015-07-30)
Medulloblastoma (MB) is composed of four molecular subgroups viz. WNT, SHH, groups 3 and 4, identified using various high-throughput methods. Translation of this molecular data into pathologist-friendly techniques that would be applicable in laboratories all over the world is a
Andreas Schmid et al.
Molecular and cellular endocrinology, 473, 225-234 (2018-02-08)
Natriuretic peptides (NPs) and their receptors gain attention regarding adipocyte function. It was the aim to investigate the expression of natriuretic peptide receptors NPR-A, NPR-B and NPR-C during adipocyte differentiation (AD), upon stimulation with fatty acids (FA), and in murine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service