Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA030620

Sigma-Aldrich

Anti-TEX14 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C20orf193, Anti-MRP-S13, Anti-MRP-S26, Anti-RPMS13, Anti-dJ534B8.3, Anti-testis expressed 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

immunogen sequence

LHFQGFIHRSLSSYAVHIISPGEARLTNLEYMLESEDRGVQRDLTRVPLPTQLYNWAAPEVILQKAATVKSDIYSFSMIMQEILTDDIPWKGLDGSVVKKAVVSGNYLEADVRLPKPYYDIVKSGIHVKQKDRTMN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TEX14(56155)

General description

TEX14 (testis-expressed sequence 14) gene is mapped in human chromosome 17q22. The encoded protein localizes in the germ cell intercellular bridge.

Immunogen

testis expressed 14 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The protein encoded by the gene TEX14 (testis-expressed sequence 14) is associated with the formation of intercellular bridges and inhibits the abscission of germ cells during cytokinesis. The intercellular bridge is formed by the interaction between Tex14 and the MKLP1 (mitotic kinesin-like protein 1)-containing midbody protein complex. TEX14 is involved in spermatogenesis and male fertility. Deletion of TEX14 interrupts intercellular bridges in all germ cells and causes male sterility, also leads to impaired kinetochore-microtubules attachment, chromosome congression defects, and whole-chromosome instability. TEX14 is an important component of mitotic kinetochore. Plk1 (Polo-like kinase 1) recruits TEX14 to kinetochore by interacting with its PBD (polo-box binding domain) domain. TEX14 regulates the maturation of the outer kinetochore and its attachment with microtubules during mitosis. The gene might be associated with testicular germ cell tumors.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75597

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and biochemical insights into the role of testis-expressed gene 14 (TEX14) in forming the stable intercellular bridges of germ cells.
Kim HJ
Proceedings of the National Academy of Sciences of the USA, 112(40), 12372-12377 (2015)
TEX14 is essential for intercellular bridges and fertility in male mice.
Greenbaum MP
Proceedings of the National Academy of Sciences of the USA, 103(13), 4982-4987 (2006)
Tokuko Iwamori et al.
Molecular and cellular biology, 30(9), 2280-2292 (2010-02-24)
In somatic cells, abscission, the physical separation of daughter cells at the completion of cytokinesis, requires CEP55, ALIX, and TSG101. In contrast, cytokinesis is arrested prior to abscission in differentiating male germ cells that are interconnected by TEX14-positive intercellular bridges.
Gourish Mondal et al.
Molecular cell, 45(5), 680-695 (2012-03-13)
Proper assembly of kinetochores (KTs) during mitosis is required for bipolar attachment of spindle microtubules (MTs) and the accumulation of spindle assembly checkpoint (SAC) components. Here we show that testis-expressed protein 14 (Tex14), which has been implicated in midbody function
K Litchfield et al.
Andrology, 3(1), 34-46 (2015-01-20)
Testicular germ cell tumour (TGCT) is the most common cause of cancer in young men (aged 15-45 years) in many populations. Multiple genome-wide association studies (GWAS) of TGCT have now been conducted, yielding over 25 disease-associated single-nucleotide polymorphism (SNP)s at 19

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service