Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA029432

Sigma-Aldrich

Anti-CLEC4C antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BDCA2, Anti-C-type lectin domain family 4, member C, Anti-CD303, Anti-CLECSF11, Anti-CLECSF7, Anti-DLEC, Anti-HECL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

MYSKTVKRLSKLREYQQYHPSLTCVMEGKDIEDWSCCPTPWTSFQSSCYFISTGMQSWTKSQKNCSVMGADLVVINTREEQDFIIQNLKRNSSYFLGLSDPGGRRHWQWVDQTPYNENVTFWHSGEPNNLDERCAIINFRSSEEWGWN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLEC4C(170482)

General description

The type II C-type lectin (CLEC4C), a transmembrane protein, is expressed on plasmacytoid dendritic cells (PDCs). CLEC4C is also known as CD303 or BDCA2 (blood dendritic cell antigen 2). This gene has seven exons and is mapped to human chromosome 12p13.31. CLEC4C codes for a member that belongs the C-type lectin (CLEC) domain family with 213 residues. The CLR, blood dendritic cell antigen 2 (BDCA2, CD303 or CLEC4C), is expressed principally on PDCs in humans.

Immunogen

C-type lectin domain family 4, member C recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CLEC4C antibody produced in rabbit has been used in immunostaining.

Biochem/physiol Actions

The type II C-type lectin (CLEC4C) plays a major role in motor neuronal vulnerability. It also participates in Ag capture, internalization and presentation to T cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72394

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CLEC4C p. K210del variant causes impaired cell surface transport in plasmacytoid dendritic cells of amyotrophic lateral sclerosis.
Lim S M, et al.
Oncotarget, 7(18), 24942-24942 (2016)
Detection of Minimal Bone Marrow involvement of Blastic Plasmacytoid Dendritic Cell Neoplastic Cells-CD303 immunostaining as a diagnostic tool.
Ohe R, et al.
Journal of clinical and experimental hepatology, 58(1), 1-9 (2018)
Targeting antigens through blood dendritic cell antigen 2 on plasmacytoid dendritic cells promotes immunologic tolerance.
Chappell CP, et al.
Journal of Immunology, 192(12), 5789?5801-5789?5801 (2014)
Lisa M Arkin et al.
iScience, 27(8), 110525-110525 (2024-08-19)
Elevated pernio incidence was observed during the COVID-19 pandemic. This prospective study enrolled subjects with pandemic-associated pernio in Wisconsin and Switzerland. Because pernio is a cutaneous manifestation of the interferonopathies, and type I interferon (IFN-I) immunity is critical to COVID-19
Rintaro Ohe et al.
Journal of clinical and experimental hematopathology : JCEH, 58(1), 1-9 (2018-02-09)
Blastic plasmacytoid dendritic cell (pDC) neoplasm (BPDCN) is a relatively rare hematological malignancy with significantly complex clinicopathological features that are still unclear. This study aimed to analyze the clinicopathological data of BPDCN and evaluate immunohistochemical detection of minimal bone marrow

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service