Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA027572

Sigma-Aldrich

Anti-EPO antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EP, Anti-erythropoietin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

APPRLICDSRVLERYLLEAKEAENITTGCAEHCSLNENITVPD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPO(2056)

General description

The gene EPO (erythropoietin) is mapped to human chromosome 7q22. In fetal cells, EPO protein is generated by the liver cells and in adults, it is produced mainly by the kidney cells. EPO is a low-molecular-weight glycoprotein hormone and a hypoxia responsive cytokine.[1]

Immunogen

erythropoietin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-EPO antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

EPO (erythropoietin) is mainly involved in erythropoiesis (controls red blood cell volume), but also has role in angiogenesis and tissue protection. Its concentration increases in conditions, such as anemia, hypoxia, renal hypoperfusion or EPO-associated hepatoprotective signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78083.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Case of Erythropoietin Producing a Large Hepatocellular Carcinoma
with Advanced Erythrocytosis
Irei T, et al.
The Japanese Society of Gastroenterological Surgery, 47, 18-18 (2014)
Chromosome 7 long arm deletion in myeloid disorders: a narrow breakpoint region in 7q22 defined by molecular mapping.
Kere J, et al.
Blood, 73, 230-234 (1989)
Hepatic erythropoietin response in cirrhosis. A contemporary review.
Ris?r LM, et al.
Scandinavian Journal of Clinical and Laboratory Investigation, 76, 183-189 (2016)
Hepatic erythropoietin response in cirrhosis.
Ris?r LM, et al.
Scandinavian Journal of Clinical and Laboratory Investigation, 76, 234-234 (2016)
Zhihong Yang et al.
International journal of clinical and experimental pathology, 8(4), 4033-4043 (2015-06-23)
To evaluate erythropoietin (Epo) and erythropoietin receptor (EpoR) expression, its relationship with vasculogenic mimicry (VM) and its prognostic value in human hepatocellular carcinoma (HCC), we examined Epo/EpoR expression and VM formation using immunohistochemistry and CD31/PAS (periodic acid-Schiff) double staining on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service