Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA026916

Sigma-Aldrich

Anti-FXYD7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FXYD domain-containing ion transport regulator 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

KKVKCRKADSRSESPTCKSCKSELPSSAPGGGGV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FXYD7(53822)

General description

FXYD domain containing ion transport regulator 7 is a member of the family of FXYD proteins, which are a group of short single- span transmembrane proteins which regulates (Na+)/(K+)ATPase. FXYD7 is a type 1 membrane protein containing N terminal that stabilises protein by post translational modifications of threonine, mainly O-glycosylations. It is expressed both in brain and astrocytes. FXYD7 is located on human chromosome 19q13.12.

Immunogen

FXYD domain-containing ion transport regulator 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FXYD7 regulates alpha 1-beta isozymes in brain.Valine residue of FXYD7 helps in endoplasmic reticulum export of FXYD7, which regulates Na,K-ATPase.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72016

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular trafficking of FXYD1 (phospholemman) and FXYD7 proteins in Xenopus oocytes and mammalian cells
Moshitzky, et al.
The Journal of Biological Chemistry, 287(25), 21130-21141 (2012)
FXYD7: Mapping of functional sites involved in ER export, association with and regulation of Na, K-ATPase
Crambert, et al.
Test (2004)
FXYD7 is a brain-specific regulator of Na, K-ATPase alpha1-beta isozymes
Beguin P,, et al.
The Embo Journal, 21(13), 3264-3273 (2002)
FXYD3 (FXYD domain containing ion transport regulator 3)
Yamamoto, et al.
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2012)
Haohao Wu et al.
Nature communications, 12(1), 1026-1026 (2021-02-17)
Proprioceptive neurons (PNs) are essential for the proper execution of all our movements by providing muscle sensory feedback to the central motor network. Here, using deep single cell RNAseq of adult PNs coupled with virus and genetic tracings, we molecularly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service