Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA026878

Sigma-Aldrich

Anti-FAT3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FAT tumor suppressor homolog 3, Anti-Protocadherin Fat 3, Anti-hFat3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

CLDSVILNNNELPLQNKRSSFAEVVGLTELKLGCVLYPDACKRSPCQHGGSCTGLPSGGYQCTCLSQFTGRNCESEITACFPNPCRNGGSCDPIGNTFICNCKAGLTG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FAT3(120114)

General description

The FAT atypical cadherin 3 (fat3) gene with 26 exons is mapped to human chromosome 11q14.3-q21. The gene codes for a 4557 amino acid protein, belonging to cadherin superfamily. Fat3 is characterized with an extracellular 33 Cadherin repeats, one Laminin G (LamG) domain and two epidermal growth factor (EGF) motifs. The encoded protein is a member of cadherin superfamily. FAT3 transcript is mainly present in embryonic stem cells (ES cells), prostate, primitive neuroectoderm, adult neural tissues, fetal brain, infant brain and adult neural tissues.

Immunogen

Protocadherin Fat 3 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FAT atypical cadherin 3 (fat3) plays a crucial role in axon fasciculation and regulation of the extracellular space encircling axons during embryonic development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72160.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mammalian fat3: a large protein that contains multiple cadherin and EGF-like motifs.
Mitsui K
Biochemical and Biophysical Research Communications, 290(4), 1260-1266 (2002)
Comparative integromics on FAT1, FAT2, FAT3 and FAT4.
Katoh Y, Katoh M
International Journal of Molecular Medicine, 18(3), 523-528 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service