Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA025064

Sigma-Aldrich

Anti-PEBP4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AC105046.10 antibody produced in rabbit, Anti-PEBP-4, Anti-Phosphatidylethanolamine-binding protein 4, Anti-Protein cousin-of-RKIP 1, Anti-hPEBP4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

EGKVISLLPKENKTRGSWKMDRFLNRFHLGEPEASTQFMTQNYQDSPTLQAPRERASEPKHK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PEBP4(157310)

General description

The gene PEBP4 (phosphatidylethanolamine binding protein 4) is mapped to human chromosome 8p21.3. It belongs to the PEBP family of proteins. The protein has potential glycosylation sites and can be secreted. It is mainly expressed in the skeletal muscle, heart and thyroid.

Immunogen

Phosphatidylethanolamine-binding protein 4 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

PEBP4 (phosphatidylethanolamine binding protein 4) regulates apoptosis by associating with Raf1 (proto-oncogene serine/threonine-protein kinase) and MEK1 (MAPK/ERK kinase 1). This association blocks TNFα (tumor necrosis α) or TRAIL (tumor necrosis factor-related apoptosis-inducing ligand) - mediated activation of MEK/ERK (extracellular signal-regulated kinase). PEBP4 is also involved in myoblast differentiation. Additionally, it promotes Akt (RAC-α serine/threonine-protein kinase) activation. The gene is expressed in various cancers and is associated with cancer progression. The PEBP4 mRNA is a target of microRNA miR-34a.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76250

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Chushi et al.
Gene, 587(1), 42-47 (2016-04-18)
Alteration of metabolic profile is one of the hallmarks of cancer cells. Statin, the inhibitors for synthesis of cholesterol, has shown anti-cancer effects on the gastric cancer cells. However, the functions of its target, HMGCR, in the progression of gastric
Il-Jin Kim et al.
Nature communications, 4, 1701-1701 (2013-04-18)
Analysis of gene expression patterns in normal tissues and their perturbations in tumours can help to identify the functional roles of oncogenes or tumour suppressors and identify potential new therapeutic targets. Here, gene expression correlation networks were derived from 92
Huan He et al.
Biochimica et biophysica acta, 1863(7 Pt A), 1682-1689 (2016-04-02)
Phosphatidylethanolamine binding proteins (PEBP) represent a superfamily of proteins that are conserved from bacteria to humans. In mammals, four members have been identified, PEBP1-4. To determine the functional differences among PEBP1-4 and the underlying mechanism for their actions, we performed
Guiping Yu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(10), 10341-10349 (2014-07-21)
The aim of this study was to determine the relationship and underlying mechanisms between ectopic expression of phosphatidylethanolamine-binding protein 4 (PEBP4) and cisplatin (DDP)-induced cytotoxicity in the lung cancer cell line A549 to provide an experimental basis for future chemotherapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service