Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA023269

Sigma-Aldrich

Anti-NIPA1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Non-imprinted in Prader-Willi/Angelman syndrome region protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

HGPTNIMVYISICSLLGSFTVPSTKGIGLAAQDILHNNPSSQRA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NIPA1(123606)

General description

The gene NIPA1 (non-imprinted in Prader-Willi/Angelman syndrome 1) is mapped to human chromosome 15q11. The protein is predicted to have nine transmembrane segments. It is widely expressed with strong expression in the brain.

Immunogen

Non-imprinted in Prader-Willi/Angelman syndrome region protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NIPA1 (non-imprinted in Prader-Willi/Angelman syndrome 1) might be involved in intracellular magnesium transport. In Drosophila, NIPA1 homolog spichthyin participates in bone morphogenic protein (BMP)-linked signaling pathways and is needed for synaptic development and axonal maintenance. Mutations in NIPA1 are associated with hereditary spastic paraplegia. NIPA1 polyalanine repeat expansions result in amyotrophic lateral sclerosis (ALS) pathogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70271

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pure hereditary spastic paraplegia due to a de novo mutation in the NIPA1 gene.
D Arkadir et al.
European journal of neurology, 21(1), e2-e2 (2014-08-19)
Jiali Zhao et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(51), 13938-13951 (2008-12-19)
We studied the consequences of expression of wild-type (WT) human NIPA1 and two mutant forms of NIPA1 with known HSP-associated mutations (T45R and G106R) on cultured rat cortical neurons and using equivalent substitutions in the Caenorhabditis elegans NIPA1 homolog CeNIPA.
Hylke M Blauw et al.
Human molecular genetics, 19(20), 4091-4099 (2010-08-06)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disease selectively affecting motor neurons in the brain and spinal cord. Recent genome-wide association studies (GWASs) have identified several common variants which increase disease susceptibility. In contrast, rare copy-number variants (CNVs), which
Shirley Rainier et al.
American journal of human genetics, 73(4), 967-971 (2003-09-26)
The hereditary spastic paraplegias (HSPs) are genetically heterogeneous disorders characterized by progressive lower-extremity weakness and spasticity. The molecular pathogenesis is poorly understood. We report discovery of a dominant negative mutation in the NIPA1 gene in a kindred with autosomal dominant
Hylke M Blauw et al.
Human molecular genetics, 21(11), 2497-2502 (2012-03-02)
Mutations in NIPA1 cause Hereditary Spastic Paraplegia type 6, a neurodegenerative disease characterized by an (upper) motor neuron phenotype. Deletions of NIPA1 have been associated with a higher susceptibility to amyotrophic lateral sclerosis (ALS). The exact role of genetic variation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service