Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA022999

Sigma-Aldrich

Anti-NBR1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-AC109326.11 antibody produced in rabbit, Anti-NBR1, Anti-neighbor of BRCA1 gene 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YSTPRLPAALEQVRLQKQVDKNFLKAEKQRLRAEKKQRKAEVKELKKQLKLHRKIHLWNSIHGLQSPKSPLGRPESLLQSNTLMLPLQPCTSVMPMLS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NBR1(4077)

General description

NBR1 (neighbor of BRCA1 gene 1) contains PB1 (Phox and Bem1), ZZ (zinc-finger domain), LIR (LC3-interacting region) and UBA (ubiquitin-associated) domains. The gene is mapped to human chromosome 17q21.

Immunogen

neighbor of BRCA1 gene 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NBR1 (neighbor of BRCA1 gene 1) is responsible for bringing ubiquitinated proteins to autophagosomes. It is also involved in the formation of protein aggregates. NBR1 works as a scaffold protein and participates in lysosomal degradation of activated FGF (fibroblast growth factor) receptors. It associates with TTN (titin) and controls SRF (serum response factor)-dependent muscle gene expression.[1] It also targets peroxisomes for pexophagy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75959

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth Deosaran et al.
Journal of cell science, 126(Pt 4), 939-952 (2012-12-15)
Selective macro-autophagy is an intracellular process by which large cytoplasmic materials are selectively sequestered and degraded in the lysosomes. Substrate selection is mediated by ubiquitylation and recruitment of ubiquitin-binding autophagic receptors such as p62, NBR1, NDP52 and Optineurin. Although it
X Liu et al.
American journal of human genetics, 64(5), 1427-1439 (1999-04-17)
Characterization of associations between polymorphic sites located throughout the approximately 200-400-kb variable-length region spanning RNU2-BRCA1 reveals nearly complete linkage disequilibrium. This segment spans the RNU2 array, which includes 6-30 tandem copies of the U2 snRNA gene, and an adjacent region
J Shi et al.
Cell death and differentiation, 21(9), 1432-1441 (2014-04-29)
Coxsackievirus infection induces an abnormal accumulation of ubiquitin aggregates that are generally believed to be noxious to the cells and have a key role in viral pathogenesis. Selective autophagy mediated by autophagy adaptor proteins, including sequestosome 1 (SQSTM1/p62) and neighbor
Anne-Sophie Nicot et al.
Autophagy, 10(6), 1036-1053 (2014-06-01)
The autophagy receptor NBR1 (neighbor of BRCA1 gene 1) binds UB/ubiquitin and the autophagosome-conjugated MAP1LC3/LC3 (microtubule-associated protein 1 light chain 3) proteins, thereby ensuring ubiquitinated protein degradation. Numerous neurodegenerative and neuromuscular diseases are associated with inappropriate aggregation of ubiquitinated proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service