Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA022539

Sigma-Aldrich

Anti-PECR antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-2,4-dienoyl-CoA reductase-related protein, Anti-DCR-RP, Anti-HPDHase, Anti-Peroxisomal trans-2-enoyl-CoA reductase, Anti-TERP, Anti-pVI-ARL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EEEVNNLVKSTLDTFGKINFLVNNGGGQFLSPAEHISSKGWHAVLETNLTGTFYMCKAVYSSWMKEHGGSIVNIIVPTKAGFPLAVHSGAARAGVYNLTKSLA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PECR(55825)

General description

Peroxisomal trans-2- enoyl-CoA reductase (PECR) is an enzyme located on the chromosome region 2q35. The carboxyl end contains PTS-1 (Peroxisomal Targeting Signal Type 1) and AKL. It has a molecular mass of 32.5kDa. The protein is highly expressed in mouse liver and kidney.

Immunogen

Peroxisomal trans-2-enoyl-CoA reductase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Peroxisomal trans-2- enoyl-CoA reductase (PECR) is involved in phytol metabolism and peroxisomal chain elongation pathway. It converts phytenoyl-CoA to phytanoyl-CoA. It is associated with fatty acid chain elongation and requires trans-2-enoyl-CoA esters for its activity. In kidney transplantation, the pre-transplant serum antibodies against PECR helps in forecasting the condition of the patient.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75476

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetics of alcohol dependence
KImura M and Higuchi S
Psychiatry and Clinical Neurosciences, 65(3), 213-225 (2011)
J Gloerich et al.
FEBS letters, 580(8), 2092-2096 (2006-03-21)
Phytol is a naturally occurring precursor of phytanic acid. The last step in the conversion of phytol to phytanoyl-CoA is the reduction of phytenoyl-CoA mediated by an, as yet, unidentified enzyme. A candidate for this reaction is a previously described
Rajani Dinavahi et al.
Journal of the American Society of Nephrology : JASN, 22(6), 1168-1178 (2011-05-14)
Although T and B cell alloimmunity contribute to transplant injury, autoimmunity directed at kidney-expressed, non-HLA antigens may also participate. Because the specificity, prevalence, and importance of antibodies to non-HLA antigens in late allograft injury are poorly characterized, we used a
A K Das et al.
The Journal of biological chemistry, 275(32), 24333-24340 (2000-05-16)
Chain elongation of fatty acids is an important cellular process and is believed to occur in the endoplasmic reticulum of all eukaroytic cells. Herein we describe the cloning and characterization of a peroxisomal NADPH-specific trans-2-enoyl-CoA reductase, the key enzyme for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service