Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA022255

Sigma-Aldrich

Anti-ZNF276 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-CENP-Z, Anti-CENPZ, Anti-MGC45417, Anti-ZADT, Anti-ZFP276, Anti-ZNF477

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PWDKETAPRLPQHRGWNPGDAPQTSQGRGTGTPVGAETKTLPSTDVAQPPSDSDAVGPRSGFPPQPSLPLCRAPGQLGEKQLPSSTSDDR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF276(92822)

General description

The gene ZNF276 (zinc finger protein 276) is mapped to human chromosome 16q24.3. It is predicted to have a miR-1226 binding motif. The protein contains C2H2 zinc fingers.

Immunogen

Zinc finger protein 276 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZNF276 (zinc finger protein 276) is a transcription factor characterized by the presence of Krüppel-associated boxes or KRAB domain. Zinc finger proteins have diverse functions that include activation of transcription, regulation of apoptosis, protein folding, RNA packaging and lipid binding. Loss of heterozygosity in ZNF276 genes has been reported in breast cancer. In genome wide association studies (GWAS), single nucleotide polymorphism in ZNF276 is associated with red hair color.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76026

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meg R Gerstenblith et al.
Pigment cell & melanoma research, 23(5), 587-606 (2010-06-16)
Recent genome-wide association studies (GWAS) identified genetic loci associated with pigmentation, nevi, and skin cancer. We performed a review and meta-analysis of GWAS results, grouping them into four categories: (i) loci associated with pigmentation (hair, eye, and/or skin color), cutaneous
Ho-Ryun Chung et al.
Molecular biology and evolution, 24(9), 1934-1943 (2007-06-16)
The zinc finger associated domain (ZAD), present in almost 100 distinct proteins, characterizes the largest subgroup of C2H2 zinc finger proteins in Drosophila melanogaster and was initially found to be encoded by arthropod genomes only. Here, we report that the
Caining Jin et al.
International journal of oncology, 37(1), 61-69 (2010-06-02)
The MUC1 oncoprotein is aberrantly overexpressed in human carcinomas and hematologic malignancies. Micro-RNAs (miRNAs) have been implicated in the suppression and induction of oncogenesis. The present studies demonstrate that the MUC1 mRNA 3' untranslated region (3'UTR) contains a highly conserved
Jasmine C Y Wong et al.
Journal of human genetics, 48(12), 668-671 (2003-11-08)
Loss of heterozygosity (LOH) involving chromosome 16q23.4 occurs frequently in breast tumors, which suggests that this region may contain a tumor suppressor gene. Since ZFP276 is located in this region, we have therefore cloned and performed mutation analysis of its
C D Constantinou-Deltas et al.
Genomics, 12(3), 581-589 (1992-03-01)
The zinc finger motif is a highly conserved tandemly repeated sequence of 28-30 amino acids that was first identified in transcription factor TFIIIA from Xenopus laevis. Subsequently, similar motifs were found and characterized in many genes from mammalian genomes and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service