Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA021775

Sigma-Aldrich

Anti-CA3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-CA-III, Anti-Carbonate dehydratase III, Anti-Carbonic anhydrase 3, Anti-Carbonic anhydrase III

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

DHWHELFPNAKGENQSPVELHTKDIRHDPSLQPWSVSYDGGSAKTILNNGKTCRVVFDDTYDRSMLRGGPLPGPYR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CA3(761)

General description

Carbonic anhydrase III (CA3) is a member of a multi-gene CA family that encodes carbonic anhydrase isozymes and termed as Car3 and CAIII. The expression of isoform has shown an increased abundance during muscle aging and is relatively muscle specific. It exhibits a fibre type-specific expression pattern.

Immunogen

Carbonic anhydrase 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-CA3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Carbonic anhydrase III (CA3) may act as a new biomarker of sarcopenia. Higher CA levels indicate an increased demand of CO2 removal in senescent muscle and the age-related fibre type shifts to slower-contracting muscles during human aging. CA3 is found to be associated with rheumatoid arthritis. CA3 possesses low carbon dioxide hydratase activity, resistance to the CA inhibitor acetazolamide unlike other members of the CA family.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74712

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huei-Yue Dai et al.
Molecular carcinogenesis, 47(12), 956-963 (2008-04-30)
Carbonic anhydrase III (CAIII) is distinguished from the other members of the CA family by low carbon dioxide hydratase activity, resistance to the CA inhibitor acetazolamide, and a predominant expression in the liver of males. In this report the effects
Lisa Staunton et al.
International journal of molecular medicine, 30(4), 723-733 (2012-07-17)
The age-related loss of skeletal muscle mass and associated progressive decline in contractile strength is a serious pathophysiological issue in the elderly. In order to investigate global changes in the skeletal muscle proteome after the fifth decade of life, this
Ai-Lian Du et al.
Autoimmunity, 42(3), 209-215 (2009-03-21)
Myasthenia gravis (MG) is considered as an autoimmune disease mainly mediated by antibodies against acetylcholine receptor. In recent years, other targets related to MG have been the subject of interest. Our previous research found that protein P25 was lower in
Julie Bachmann et al.
PLoS pathogens, 10(4), e1004038-e1004038 (2014-04-20)
Systemic inflammation and sequestration of parasitized erythrocytes are central processes in the pathophysiology of severe Plasmodium falciparum childhood malaria. However, it is still not understood why some children are more at risks to develop malaria complications than others. To identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service