Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA021430

Sigma-Aldrich

Anti-SPECC1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-CYTSB, Anti-NSP5, Anti-Nuclear structure protein 5, Anti-Sperm antigen HCMOGT-1, Anti-Sperm antigen with calponin homology and coiled-coil domains 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

APREFSVTVSRERSVPRGPSNPRKSVSSPTSSNTPTPTKHLRTPSTKPKQENEGGEKAALESQVRELLAEAKAKDSEINRLRSELKKYKEKRTLNA

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYTSB(92521)

General description

The gene SPECC1 (sperm antigen with calponin homology and coiled-coil domains 1) is mapped to human chromosome 17p11.2. It is mainly expressed in testis and localizes in the nucleus. SPECC1 is commonly referred as NSP5 (nuclear structure protein 5).

Immunogen

Sperm antigen with calponin homology and coiled-coil domains 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SPECC1 (sperm antigen with calponin homology and coiled-coil domains 1) is up-regulated in certain cancer cells. It mediates apoptosis in head and neck carcinoma. SPECC1 associates with B23 (nucleophosmin) and ribonuclear protein HNRNPL (heterogeneous nuclear ribonucleoprotein L), suggesting its role in ribosome biogenesis and rRNA transcription. Chromosomal translocation in juvenile myelomonocytic leukemia results in SPECC1-PDGFRB (platelet-derived growth factor receptor β) fusion protein.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75594

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nianli Sang et al.
Oncogene, 23(58), 9438-9446 (2004-12-17)
We isolated several related but distinct cDNA clones encoding novel structure proteins (NSP) when screening a cDNA library. Analysis revealed that these cDNAs and several similar ESTs in the public databases are derived from a single gene of 17 exons
Luca D'Agostino et al.
Oncotarget, 2(12), 1055-1074 (2011-12-16)
NSP 5a3a is a novel structural protein found to be over-expressed in certain cancer cell lines in-vitro such as Hela, Saos-2, and MCF-7 while barely detectable levels in normal body tissues except for Testis. This particular isoform has been known
Cristina Morerio et al.
Cancer research, 64(8), 2649-2651 (2004-04-17)
PDGFRB, a transmembrane tyrosine kinase receptor for platelet-derived growth factor, is constitutively activated by gene fusion with different partners in myeloproliferative/myelodysplastic disorders with peculiar clinical characteristics. Six alternative partner genes have been described thus far. In this study, we report
Luca D'Agostino et al.
Cell cycle (Georgetown, Tex.), 9(6), 1131-1142 (2010-03-20)
NSP 5a3a had been identified along with three other distinct though similar isoforms corresponding to locus HCMOGT-1 on chromosome 17p11.2. ( 1) Secondary structure analysis of the NSP isoforms revealed similarity to Spectrin and Spectrin like proteins containing coiled coil

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service